Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Parp3
Entrez Gene ID
235587
Gene Name
poly (ADP-ribose) polymerase family, member 3
Gene Aliases
A930002C11Rik, AW990611, Adprt3, Adprtl3, PARP-3, pADPRT-3
Chromosome Location
Chr.9: 106470353 - 106476944 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145619.2 NP_663594.2 8 - 9 1243 62
GenBank mRNA AF368233.1 - 8 - 9 1243 62
AK044223.1 - 8 - 9 1236 62
AK088125.1 - 8 - 9 1249 62
AK145259.1 - 8 - 9 1277 62
AK147868.1 - 8 - 9 1542 62
AK170541.1 - 8 - 9 1187 62
BC014870.1 - 8 - 9 1183 62
BC058754.1 - 8 - 9 1163 62
BC062204.1 - 8 - 9 1163 62

View Full Product Details