Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ercc1
Entrez Gene ID
13870
Gene Name
excision repair cross-complementing rodent repair deficiency, complementation group 1
Gene Aliases
Ercc-1
Chromosome Location
Chr.7: 19344422 - 19356524 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127324.1 NP_001120796.1 4 - 5 559 58
NM_007948.2 NP_031974.2 4 - 5 559 58
GenBank mRNA AK088271.1 - 4 - 5 531 58
AK136733.1 - 4 - 5 519 58
AK160703.1 - 4 - 5 482 58
BC011224.1 - 4 - 5 529 58
X07414.1 - 4 - 5 524 58

View Full Product Details