Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tinagl1
Entrez Gene ID
94242
Gene Name
tubulointerstitial nephritis antigen-like 1
Gene Aliases
1110021J17Rik, AZ-1, AZ1, Arg1, Lcn7, TARP, Tinagl
Chromosome Location
Chr.4: 130165600 - 130175122 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168333.1 NP_001161805.1 2 - 3 391 82
NM_023476.3 NP_075965.2 2 - 3 532 82
GenBank mRNA AB047402.1 - 2 - 3 533 82
AB050626.1 - 2 - 3 369 82
AK087142.1 - - 391 82
BC005738.1 - 2 - 3 359 82
BC018539.1 - 2 - 3 376 82
CT010275.1 - 1 - 2 150 82

View Full Product Details