Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sytl2
Entrez Gene ID
83671
Gene Name
synaptotagmin-like 2
Gene Aliases
AI266830, Slp2, mKIAA1597
Chromosome Location
Chr.7: 90302207 - 90410719 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040085.2 NP_001035174.1 15 - 16 3032 106
NM_001040087.2 NP_001035176.1 6 - 7 1239 106
NM_001040088.2 NP_001035177.1 6 - 7 1167 106
NM_001289583.1 NP_001276512.1 16 - 17 3080 106
NM_001289584.1 NP_001276513.1 16 - 17 2999 106
NM_001289585.1 NP_001276514.1 7 - 8 1332 106
NM_001289586.1 NP_001276515.1 7 - 8 1287 106
NM_031394.3 NP_113571.2 15 - 16 2960 106
GenBank mRNA AB050742.1 - 6 - 7 778 106
AB057754.1 - 15 - 16 2419 106
AB057755.1 - 9 - 10 2929 106
AB057756.1 - 7 - 8 1254 106
AB057757.1 - 2 - 3 1245 106
AB057760.1 - 15 - 16 2338 106
AB057761.1 - 14 - 15 2371 106
AB057762.1 - 14 - 15 2299 106
AB057763.1 - 3 - 4 1148 106
AK027924.1 - 6 - 7 1150 106
AK051653.1 - 7 - 8 1333 106
AK173212.1 - 6 - 7 697 106
BC060665.1 - 7 - 8 2782 106
BC138714.1 - 14 - 15 2270 106
BC145398.1 - 15 - 16 2390 106

View Full Product Details