The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289732.1 | NP_001276661.1 | 4 - 5 | 483 | 75 |
NM_001289733.1 | NP_001276662.1 | 3 - 4 | 401 | 75 | |
NM_033605.2 | NP_291083.1 | 2 - 3 | 723 | 75 | |
GenBank mRNA | AF257217.1 | - | 2 - 3 | 722 | 75 |
AK033050.1 | - | 4 - 5 | 484 | 75 | |
AK041446.1 | - | 3 - 4 | 390 | 75 | |
AK045154.1 | - | 2 - 3 | 726 | 75 |