Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alppl2
Entrez Gene ID
11650
Gene Name
alkaline phosphatase, placental-like 2
Gene Aliases
Akp5, C77216, D1Ertd816e, EAP
Chromosome Location
Chr.1: 87086690 - 87096250 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007433.3 NP_031459.3 6 - 7 947 59
GenBank mRNA AK145571.1 - 7 - 8 856 59
AY054302.1 - 6 - 7 788 59
BC052874.1 - 6 - 7 942 59

View Full Product Details