Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfs1
Entrez Gene ID
18041
Gene Name
nitrogen fixation gene 1 (S. cerevisiae)
Gene Aliases
AA987187, m-Nfs1, m-Nfsl
Chromosome Location
Chr.2: 156123637 - 156144186 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010911.2 NP_035041.2 5 - 6 615 62
GenBank mRNA AJ222660.1 - 5 - 6 614 62
AK005724.1 - 5 - 6 604 62
AK075575.1 - 5 - 6 613 62
AK132838.1 - 5 - 6 616 62
AK148527.1 - 5 - 6 616 62
AK159828.1 - 5 - 6 604 62
BC010586.1 - 5 - 6 593 62

View Full Product Details