Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Numb
Entrez Gene ID
18222
Gene Name
numb gene homolog (Drosophila)
Gene Aliases
Nb
Chromosome Location
Chr.12: 83794034 - 83921934 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136075.2 NP_001129547.1 2 - 3 211 101
NM_010949.2 NP_035079.1 2 - 3 211 101
NR_073564.1 5 - 6 558 101
GenBank mRNA AF169192.1 - 2 - 3 196 101
AF170709.1 - 2 - 3 196 101
BC050108.1 - 5 - 6 468 101

View Full Product Details