The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136075.2 | NP_001129547.1 | 5 - 6 | 462 | 62 |
NM_001272055.1 | NP_001258984.1 | 4 - 5 | 429 | 62 | |
NM_001272056.1 | NP_001258985.1 | 6 - 7 | 659 | 62 | |
NM_010949.2 | NP_035079.1 | 5 - 6 | 462 | 62 | |
NR_073563.1 | 7 - 8 | 776 | 62 | ||
NR_073564.1 | 8 - 9 | 809 | 62 | ||
GenBank mRNA | AF169191.1 | - | 4 - 5 | 414 | 62 |
AF169192.1 | - | 5 - 6 | 447 | 62 | |
AF170709.1 | - | 5 - 6 | 447 | 62 | |
AK004553.1 | - | 7 - 8 | 684 | 62 | |
BC033459.1 | - | 7 - 8 | 700 | 62 | |
BC050108.1 | - | 8 - 9 | 719 | 62 | |
U70674.1 | - | 6 - 7 | 580 | 62 |