Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptcra
Entrez Gene ID
19208
Gene Name
pre T cell antigen receptor alpha
Gene Aliases
gp33, pT-alpha, pT-alpha-TCR, pT[a], pTa, pTalpha
Chromosome Location
Chr.17: 46755663 - 46763712 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011195.2 NP_035325.1 1 - 2 179 65
GenBank mRNA BC119062.1 - 1 - 2 104 65
BC120746.1 - 1 - 2 104 65
U16958.1 - 1 - 2 179 65

View Full Product Details