Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acpp
Entrez Gene ID
56318
Gene Name
acid phosphatase, prostate
Gene Aliases
5'-NT, A030005E02Rik, FRAP, Lap, PAP, Ppal
Chromosome Location
Chr.9: 104288240 - 104337722 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019807.2 NP_062781.2 6 - 7 704 91
NM_207668.2 NP_997551.1 6 - 7 704 91
GenBank mRNA AF210243.1 - 6 - 7 657 91
AK020693.1 - 6 - 7 666 91
AK029273.1 - 6 - 7 690 91
AK076383.1 - 6 - 7 711 91
BC139826.1 - 6 - 7 695 91

View Full Product Details