The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019996.4 | NP_064380.3 | 1 - 2 | 265 | 108 |
GenBank mRNA | AJ276504.1 | - | 1 - 2 | 96 | 108 |
AK012949.1 | - | 1 - 2 | 108 | ||
AK030749.1 | - | 1 - 2 | 123 | 108 | |
BC016530.1 | - | 1 - 2 | 112 | 108 | |
BC049590.1 | - | 1 - 2 | 111 | 108 | |
BC061110.1 | - | 1 - 2 | 125 | 108 | |
CJ157832.1 | - | 1 - 2 | 266 | 108 |