The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020575.2 | NP_065600.1 | 7 - 8 | 2055 | 102 |
GenBank mRNA | AF155739.1 | - | 7 - 8 | 2055 | 102 |
AK143177.1 | - | 7 - 8 | 2077 | 102 | |
AK166666.1 | - | 7 - 8 | 2062 | 102 | |
AK167246.1 | - | 7 - 8 | 2037 | 102 | |
AK168555.1 | - | 7 - 8 | 2058 | 102 | |
BC025029.1 | - | 7 - 8 | 2019 | 102 |