Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cct2
Entrez Gene ID
12461
Gene Name
chaperonin containing Tcp1, subunit 2 (beta)
Gene Aliases
Cctb
Chromosome Location
Chr.10: 117050997 - 117063814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007636.2 NP_031662.2 10 - 11 1084 82
GenBank mRNA AB041570.1 - 10 - 11 1056 82
AK075581.1 - 10 - 11 1057 82
AK088145.1 - 10 - 11 1046 82
AK145406.1 - 10 - 11 1085 82
AK166891.1 - 10 - 11 1057 82
BC007470.1 - 10 - 11 1048 82
BC026918.1 - 10 - 11 1050 82
Z31553.1 - 10 - 11 1035 82

View Full Product Details