The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252481.1 | NP_001239410.1 | 2 - 3 | 675 | 79 |
NM_010754.5 | NP_034884.2 | 2 - 3 | 564 | 79 | |
GenBank mRNA | AF005743.1 | - | 1 - 2 | 292 | 79 |
AK007817.1 | - | 2 - 3 | 341 | 79 | |
BC021342.1 | - | 2 - 3 | 363 | 79 | |
BC089184.1 | - | 2 - 3 | 675 | 79 | |
U60530.1 | - | 1 - 2 | 292 | 79 |