Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smad2
Entrez Gene ID
17126
Gene Name
SMAD family member 2
Gene Aliases
7120426M23Rik, Madh2, Madr2, Smad-2, mMad2
Chromosome Location
Chr.18: 76241251 - 76311748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252481.1 NP_001239410.1 8 - 9 1442 80
NM_010754.5 NP_034884.2 8 - 9 1331 80
GenBank mRNA AF005743.1 - 7 - 8 1059 80
AK007817.1 - 8 - 9 1108 80
AY334552.1 - 6 - 7 910 80
BC021342.1 - 8 - 9 1130 80
BC089184.1 - 8 - 9 1442 80
U60530.1 - 7 - 8 1059 80

View Full Product Details