Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpgr
Entrez Gene ID
19893
Gene Name
retinitis pigmentosa GTPase regulator
Gene Aliases
Rp3h
Chromosome Location
Chr.X: 10158216 - 10216795 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177950.1 NP_001171421.1 16 - 17 2920 94
NM_001177951.1 NP_001171422.1 16 - 17 2920 94
NM_001177952.1 NP_001171423.1 15 - 16 2035 94
NM_011285.2 NP_035415.1 15 - 16 2041 94
GenBank mRNA AF044677.1 - 15 - 16 1959 94
AJ238396.1 - 16 - 17 2832 94
AK029864.1 - 16 - 17 3046 94
AK136298.1 - 15 - 16 1972 94
AK138977.1 - 15 - 16 2036 94
BC012661.1 - 15 - 16 1980 94

View Full Product Details