Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stk11
Entrez Gene ID
20869
Gene Name
serine/threonine kinase 11
Gene Aliases
AA408040, Lkb1, Par-4, R75140
Chromosome Location
Chr.10: 80115693 - 80130685 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB015801.1 - 1 - 2 336 87
AF129870.1 - 1 - 2 714 87
AF145287.1 - 1 - 2 318 87
AF151711.1 - 1 - 2 318 87
AK172385.1 - 1 - 2 1101 87
AK172528.1 - 1 - 2 1098 87
BC052379.1 - 1 - 2 379 87
CO424479.1 - 1 - 2 536 87
EU730638.1 - 1 - 2 297 87

View Full Product Details