The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011797.2 | NP_035927.1 | 2 - 3 | 341 | 81 |
GenBank mRNA | AB005450.1 | - | 2 - 3 | 341 | 81 |
AK157764.1 | - | 2 - 3 | 341 | 81 | |
BC046995.1 | - | 2 - 3 | 323 | 81 |