The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011885.4 | NP_036015.1 | 2 - 3 | 81 | 96 |
GenBank mRNA | AK146730.1 | - | 2 - 3 | 103 | 96 |
BC012679.1 | - | 2 - 3 | 117 | 96 | |
BC027522.1 | - | 2 - 3 | 86 | 96 | |
Y11682.1 | - | 2 - 3 | 80 | 96 |