Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fto
Entrez Gene ID
26383
Gene Name
fat mass and obesity associated
Gene Aliases
AW743446, mKIAA1752
Chromosome Location
Chr.8: 91313491 - 91668433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011936.2 NP_036066.2 1 - 2 94 63
GenBank mRNA AJ237917.1 - 1 - 2 80 63
AK036677.1 - 1 - 2 84 63
AK040866.1 - 1 - 2 72 63
AK045465.1 - 1 - 2 71 63
AK046935.1 - 1 - 2 53 63
AK049502.1 - 1 - 2 87 63
AK088881.1 - 1 - 2 80 63
AK129437.1 - 1 - 2 61 63
AK161060.1 - 1 - 2 53 63
BC022222.1 - 1 - 2 45 63
BC057008.1 - 1 - 2 60 63
BY290198.1 - 1 - 2 95 63

View Full Product Details