Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6galnac5
Entrez Gene ID
26938
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases
AI851940, Siat7e
Chromosome Location
Chr.3: 152820709 - 152982207 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012028.4 NP_036158.3 4 - 5 965 69
GenBank mRNA AB028840.1 - - 947 69
AB030836.1 - 4 - 5 968 69
AK034387.1 - - 969 69
AK038434.1 - 4 - 5 69
AK042683.1 - 4 - 5 1005 69
BC055737.1 - 4 - 5 965 69

View Full Product Details