The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013679.2 | NP_038707.2 | 1 - 2 | 98 | 113 |
GenBank mRNA | AK020303.1 | - | 1 - 2 | 100 | 113 |
BC119247.1 | - | 1 - 2 | 102 | 113 | |
BC119249.1 | - | 1 - 2 | 102 | 113 | |
L11393.1 | - | 1 - 2 | 90 | 113 | |
S65753.1 | - | 1 - 2 | 90 | 113 |