The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253877.1 | NP_001240806.1 | 8 - 9 | 1373 | 78 |
NM_001253878.1 | NP_001240807.1 | 8 - 9 | 1092 | 78 | |
NM_001253879.1 | NP_001240808.1 | 7 - 8 | 1015 | 78 | |
NM_001253880.1 | NP_001240809.1 | 5 - 6 | 730 | 78 | |
NM_013827.3 | NP_038855.2 | 7 - 8 | 1011 | 78 | |
NR_045608.1 | 9 - 10 | 1079 | 78 | ||
GenBank mRNA | AB221622.1 | - | 4 - 5 | 418 | 78 |
AK030752.1 | - | 7 - 8 | 961 | 78 | |
AK031409.1 | - | 9 - 10 | 1080 | 78 | |
AK032798.1 | - | 8 - 9 | 1334 | 78 | |
AK032819.1 | - | 8 - 9 | 1335 | 78 | |
AK137539.1 | - | 9 - 10 | 1320 | 78 | |
AK139685.1 | - | 8 - 9 | 1093 | 78 | |
BC024889.1 | - | 7 - 8 | 983 | 78 | |
BC029076.1 | - | 7 - 8 | 922 | 78 | |
BC050826.1 | - | 7 - 8 | 965 | 78 | |
BC057163.1 | - | 7 - 8 | 808 | 78 | |
BC071223.1 | - | 5 - 6 | 483 | 78 | |
BC092237.1 | - | 7 - 8 | 943 | 78 | |
BC100340.1 | - | 5 - 6 | 661 | 78 | |
S78454.1 | - | 4 - 5 | 418 | 78 |