Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl39
Entrez Gene ID
27393
Gene Name
mitochondrial ribosomal protein L39
Gene Aliases
C21orf8, MRP-L5, ORF22, Rpml5
Chromosome Location
Chr.16: 84717580 - 84735302 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017404.4 NP_059100.3 4 - 5 597 92
GenBank mRNA AF239728.1 - 4 - 5 514 92
AK007681.1 - 4 - 5 533 92
AK031942.1 - 4 - 5 518 92
BC012274.1 - 4 - 5 476 92

View Full Product Details