Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sfrp5
Entrez Gene ID
54612
Gene Name
secreted frizzled-related sequence protein 5
Gene Aliases
AI605071, SARP3
Chromosome Location
Chr.19: 42197971 - 42202252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018780.3 NP_061250.2 1 - 2 768 85
GenBank mRNA AF117759.1 - 1 - 2 768 85
BC032921.1 - 1 - 2 647 85

View Full Product Details