The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018828.2 | NP_061298.1 | 12 - 13 | 2253 | 97 |
GenBank mRNA | AK084018.1 | - | 12 - 13 | 2045 | 97 |
AK129263.1 | - | 11 - 12 | 3124 | 97 | |
U40750.2 | - | 12 - 13 | 2253 | 97 |