Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apip
Entrez Gene ID
56369
Gene Name
APAF1 interacting protein
Gene Aliases
APIP2, CGI-29, Mmrp19
Chromosome Location
Chr.2: 103073500 - 103092649 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019735.4 NP_062709.3 1 - 2 164 82
GenBank mRNA AB028863.1 - 1 - 2 56 82
AK145450.1 - 1 - 2 166 82
BC028434.1 - 1 - 2 107 82

View Full Product Details