The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019937.3 | NP_064321.2 | 2 - 3 | 491 | 93 |
GenBank mRNA | AF159159.1 | - | 2 - 3 | 444 | 93 |
AF467251.1 | - | 2 - 3 | 485 | 93 | |
AK051380.1 | - | 2 - 3 | 448 | 93 | |
BC007177.1 | - | 2 - 3 | 465 | 93 | |
BC037204.1 | - | 2 - 3 | 429 | 93 | |
BC069909.1 | - | 2 - 3 | 428 | 93 | |
BC094383.1 | - | 2 - 3 | 427 | 93 | |
BC117816.1 | - | 2 - 3 | 311 | 93 |