Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnb1
Entrez Gene ID
16500
Gene Name
potassium voltage gated channel, Shab-related subfamily, member 1
Gene Aliases
Kcr1-1, Kv2.1, Shab
Chromosome Location
Chr.2: 167095969 - 167188818 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008420.4 NP_032446.2 1 - 2 761 73
GenBank mRNA BC031776.1 - 1 - 2 717 73
BC051422.1 - 1 - 2 718 73
BC061501.1 - 1 - 2 742 73
DQ646376.1 - - 1015 73
DQ646377.1 - 1 - 2 761 73
M64228.1 - 1 - 2 566 73

View Full Product Details