The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008674.2 | NP_032700.1 | 3 - 3 | 587 | 102 |
GenBank mRNA | AK077364.1 | - | 3 - 3 | 587 | 102 |