The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010357.3 | NP_034487.2 | 6 - 7 | 670 | 87 |
GenBank mRNA | AI043064.1 | - | 1 - 2 | 87 | |
AK008189.1 | - | 6 - 7 | 656 | 87 | |
AK008193.1 | - | 7 - 8 | 621 | 87 | |
AK008400.1 | - | 6 - 7 | 654 | 87 | |
AK008490.1 | - | 6 - 7 | 621 | 87 | |
AK009668.1 | - | 6 - 7 | 624 | 87 | |
AK010098.1 | - | 6 - 7 | 624 | 87 | |
AK011177.1 | - | 6 - 7 | 621 | 87 | |
AK011841.1 | - | 6 - 7 | 649 | 87 | |
AK019271.1 | - | 6 - 7 | 645 | 87 | |
BC012639.1 | - | 6 - 7 | 624 | 87 | |
L06047.1 | - | 6 - 7 | 622 | 87 |