Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Il11ra1
- Entrez Gene ID
-
16157
- Gene Name
- interleukin 11 receptor, alpha chain 1
- Gene Aliases
-
AI314697, GP130, Il-11ra, Il11ra, Il11ra2, NR1
- Chromosome Location
-
Chr.4: 41760443 - 41769473 on Build GRCm38
- Gene Symbol
- Gm13305
- Entrez Gene ID
-
100042555
- Gene Name
- predicted gene 13305
- Gene Aliases
-
OTTMUSG00000011351
- Chromosome Location
-
Chr.4: 42158842 - 42168603 on Build GRCm38
- Gene Symbol
- Gm2002
- Entrez Gene ID
-
100038993
- Gene Name
- predicted gene 2002
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4|NT_187054: 3084 - 13572 on Build GRCm38
- Gene Symbol
- Il11ra2
- Entrez Gene ID
-
16158
- Gene Name
- interleukin 11 receptor, alpha chain 2
- Gene Aliases
-
ENSMUSG00000066201, Etl-2, Etl2, Il11ra-ps1
- Chromosome Location
-
Chr.4: 42655275 - 42665763 on Build GRCm38
View Full Product Details