Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wfs1
Entrez Gene ID
22393
Gene Name
Wolfram syndrome 1 homolog (human)
Gene Aliases
AI481085, wolframin
Chromosome Location
Chr.5: 36966104 - 36988982 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011716.2 NP_035846.1 5 - 6 754 87
GenBank mRNA AF084482.1 - 5 - 6 87
AJ011971.1 - 5 - 6 681 87
AK144561.1 - 5 - 6 706 87
AK145023.1 - 5 - 6 726 87
AK170180.1 - 5 - 6 709 87
BC046988.1 - 5 - 6 690 87
CX562372.1 - 3 - 4 384 87

View Full Product Details