Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb33
Entrez Gene ID
56805
Gene Name
zinc finger and BTB domain containing 33
Gene Aliases
AW260253, E130014G12Rik, Kaiso, Znf-kaiso
Chromosome Location
Chr.X: 38189793 - 38197046 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020256.2 NP_064652.2 1 - 2 292 83
GenBank mRNA AF097416.1 - 1 - 2 255 83
AK089423.1 - 1 - 2 252 83
BC138647.1 - 1 - 2 91 83
BC138648.1 - 1 - 2 91 83

View Full Product Details