Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap21-1
Entrez Gene ID
170656
Gene Name
keratin associated protein 21-1
Gene Aliases
1110017B05Rik, AI506979, KAP21.1, Krtap16-7
Chromosome Location
Chr.16: 89403027 - 89403774 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028621.3 NP_082897.2 1 - 1 372 117
GenBank mRNA AF345297.1 - 1 - 1 117
BC145974.1 - 1 - 1 372 117
BC145976.1 - 1 - 1 372 117
BC147776.1 - 1 - 1 466 117
BC147786.1 - 1 - 1 466 117

View Full Product Details