Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cstf2
Entrez Gene ID
108062
Gene Name
cleavage stimulation factor, 3' pre-RNA subunit 2
Gene Aliases
C630034J23Rik, Cstf64, mir-3112
Chromosome Location
Chr.X: 134059139 - 134086822 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290398.1 NP_001277327.1 7 - 8 1019 63
NM_001290399.1 NP_001277328.1 7 - 8 1019 63
NM_133196.6 NP_573459.1 7 - 8 1019 63
GenBank mRNA AF317552.1 - 7 - 8 987 63
AK032817.1 - 7 - 8 1001 63
AK049971.1 - 7 - 8 1010 63
AK088260.1 - 7 - 8 847 63
AK147438.1 - 6 - 7 63
BC036719.1 - 7 - 8 991 63
EU616680.1 - 7 - 8 987 63
EU616681.1 - 7 - 8 987 63
EU616682.1 - 7 - 8 987 63

View Full Product Details