Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pygl
Entrez Gene ID
110095
Gene Name
liver glycogen phosphorylase
Gene Aliases
-
Chromosome Location
Chr.12: 70190815 - 70227683 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133198.2 NP_573461.2 4 - 5 629 93
GenBank mRNA AF288783.1 - 4 - 5 571 93
AK140321.1 - 4 - 5 576 93
AK145989.1 - 4 - 5 630 93
AK149491.1 - - 804 93
AK167337.1 - 4 - 5 576 93
BC013636.1 - 4 - 5 563 93

View Full Product Details