The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007998.6 | NP_032024.2 | 4 - 5 | 558 | 76 |
GenBank mRNA | AK004718.2 | - | 4 - 5 | 559 | 76 |
AK018738.2 | - | 4 - 5 | 561 | 76 | |
AK084843.1 | - | 4 - 5 | 456 | 76 | |
AK150685.1 | - | 4 - 5 | 560 | 76 | |
AK159652.1 | - | 4 - 5 | 591 | 76 | |
BC006746.1 | - | 4 - 5 | 565 | 76 | |
BC094548.1 | - | 4 - 5 | 531 | 76 | |
BC096770.1 | - | 3 - 4 | 392 | 76 | |
M59288.1 | - | 3 - 4 | 395 | 76 | |
M61215.1 | - | 4 - 5 | 545 | 76 |