The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009708.1 | NP_033838.1 | 3 - 4 | 305 | 85 |
GenBank mRNA | AK075970.1 | - | - | 613 | 85 |
AK170529.1 | - | - | 1736 | 85 | |
BC107022.2 | - | 3 - 4 | 304 | 85 | |
BC127034.1 | - | 3 - 4 | 299 | 85 | |
BC128475.1 | - | 3 - 4 | 304 | 85 | |
BC139330.1 | - | 3 - 4 | 378 | 85 | |
BC139331.1 | - | 3 - 4 | 378 | 85 |