The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163161.1 | NP_001156633.1 | 5 - 6 | 720 | 89 |
NM_010819.4 | NP_034949.3 | 5 - 6 | 723 | 89 | |
GenBank mRNA | AF061272.1 | - | 5 - 6 | 655 | 89 |
AK089500.1 | - | 5 - 6 | 714 | 89 | |
AK150926.1 | - | 5 - 6 | 626 | 89 | |
AK170692.1 | - | 5 - 6 | 584 | 89 | |
AK171409.1 | - | 5 - 6 | 661 | 89 | |
BC099471.1 | - | 5 - 6 | 633 | 89 |