Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc8c
Entrez Gene ID
100604
Gene Name
leucine rich repeat containing 8 family, member C
Gene Aliases
AD158, AI326115, E430036I04Rik, fad158
Chromosome Location
Chr.5: 105519386 - 105613018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133897.2 NP_598658.1 3 - 4 371 84
GenBank mRNA AB081508.1 - 3 - 4 371 84
AK089024.1 - 3 - 4 439 84
AK142337.1 - 3 - 4 401 84
AK169492.1 - 3 - 4 434 84
AK171313.1 - 3 - 4 453 84
BC026572.1 - 3 - 4 381 84

View Full Product Details