The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133922.3 | NP_598683.3 | 3 - 4 | 558 | 90 |
GenBank mRNA | AB041543.1 | - | 2 - 3 | 391 | 90 |
AK040525.1 | - | 3 - 4 | 560 | 90 | |
AK128931.1 | - | 3 - 4 | 512 | 90 | |
AK162934.1 | - | 3 - 4 | 266 | 90 | |
AK173281.1 | - | 3 - 4 | 2262 | 90 | |
BC066816.1 | - | 3 - 4 | 478 | 90 |