Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chodl
Entrez Gene ID
246048
Gene Name
chondrolectin
Gene Aliases
3110074E07Rik, MT75, PRED12
Chromosome Location
Chr.16: 78930864 - 78951733 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139134.3 NP_624360.2 1 - 2 497 83
GenBank mRNA AF311699.1 - 1 - 2 320 83
AK031063.1 - 1 - 2 498 83
AK076523.1 - 1 - 2 497 83
AK140910.1 - 1 - 2 498 83
BC117071.1 - 1 - 2 168 83
BC117073.1 - 1 - 2 168 83
BC144738.1 - 1 - 2 168 83

View Full Product Details