Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Artn
Entrez Gene ID
11876
Gene Name
artemin
Gene Aliases
neublastin
Chromosome Location
Chr.4: 117926160 - 117930791 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284191.1 NP_001271120.1 5 - 6 927 77
NM_001284193.1 NP_001271122.1 4 - 5 1413 77
NM_009711.4 NP_033841.1 4 - 5 1269 77
GenBank mRNA AF109402.1 - 2 - 3 198 77
AK015393.1 - 5 - 6 635 77
AK053914.1 - 4 - 5 1270 77
AK156507.1 - 4 - 5 1171 77

View Full Product Details