The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025885.3 | NP_080161.1 | 1 - 2 | 259 | 87 |
GenBank mRNA | AK005399.1 | - | 1 - 2 | 227 | 87 |
AK010552.1 | - | 1 - 2 | 237 | 87 | |
AK160579.1 | - | 1 - 2 | 225 | 87 | |
AK168787.1 | - | 1 - 2 | 227 | 87 | |
BB611514.1 | - | 1 - 2 | 259 | 87 | |
BC044207.1 | - | 1 - 2 | 121 | 87 | |
BC044302.1 | - | 1 - 2 | 121 | 87 | |
BC055404.1 | - | 1 - 2 | 214 | 87 | |
BC082559.1 | - | 1 - 2 | 224 | 87 |