Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex11g
Entrez Gene ID
69129
Gene Name
peroxisomal biogenesis factor 11 gamma
Gene Aliases
1810022F11Rik, 1810049N02Rik, Pex11c, Pex11gamma
Chromosome Location
Chr.8: 3457105 - 3467839 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026951.2 NP_081227.1 1 - 2 80 96
GenBank mRNA AK007582.1 - 1 - 2 81 96
AK165461.1 - 1 - 2 102 96
BC024096.1 - 1 - 2 112 96
BC062135.1 - 1 - 2 69 96

View Full Product Details