Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbs5
Entrez Gene ID
72569
Gene Name
Bardet-Biedl syndrome 5 (human)
Gene Aliases
1700049I01Rik, 2700023J09Rik
Chromosome Location
Chr.2: 69646730 - 69667569 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028284.2 NP_082560.1 3 - 4 228 98
GenBank mRNA AK012283.1 - 3 - 4 228 98
BC061031.1 - 3 - 4 229 98

View Full Product Details