Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmco4
Entrez Gene ID
77056
Gene Name
transmembrane and coiled-coil domains 4
Gene Aliases
4632413C14Rik, AI430825
Chromosome Location
Chr.4: 138972901 - 139059173 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029857.3 NP_084133.1 7 - 8 1148 93
GenBank mRNA AK014571.1 - 6 - 7 749 93
AK029092.1 - 7 - 8 1148 93
AK054337.1 - 7 - 8 1145 93
AK170619.1 - 7 - 8 877 93
AK171189.1 - 7 - 8 894 93
BC013471.1 - 6 - 7 774 93

View Full Product Details