The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145706.2 | NP_663752.2 | 3 - 4 | 380 | 96 |
GenBank mRNA | AK090019.1 | - | 3 - 4 | 343 | 96 |
AK141339.1 | - | 3 - 4 | 345 | 96 | |
AK146262.1 | - | 3 - 4 | 382 | 96 | |
BC011060.1 | - | 3 - 4 | 368 | 96 | |
BC052530.1 | - | 3 - 4 | 308 | 96 | |
BF607996.1 | - | 2 - 3 | 148 | 96 |